Learn More
Cadmium acetate dihydrate, 98%, pure, Thermo Scientific™
Chemical Identifiers
| CAS | 5743-04-4 |
|---|---|
| Molecular Formula | C4H6CdO4·2H2O |
| Molecular Weight (g/mol) | 266.52 |
| InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC212310010
|
Thermo Scientific Chemicals
212310010 |
1kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC212310050
|
Thermo Scientific Chemicals
212310050 |
5kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5743-04-4 | |
| C4H6CdO4·2H2O | |
| 266.52 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Specifications
| 5743-04-4 | |
| 97.5% min. (Complexometry) | |
| (CH3CO2)2Cd·2H2O | |
| Solubility in water: soluble. Other solubilities: soluble in ethanol | |
| 266.52 | |
| 98% | |
| 12.5% to 14.5% |
| 1kg | |
| C4H6CdO4·2H2O | |
| 10,1584 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| 266.52 | |
| Pure | |
| Cadmium acetate dihydrate |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a posi
GHS Signal Word: Danger
RUO â Research Use Only