missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 98%, for analysis

Catalog No. AC317135000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317135000 500 g Plastic bottle
AC317130025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC317135000 Supplier Thermo Scientific Chemicals Supplier No. 317135000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H6CdO4·2H2O
Molecular Weight (g/mol) 266.52
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CH3CO2)2Cd·2H2O
Merck Index 10,1584
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 266.52
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Cadmium acetate dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a posi

RUO – Research Use Only