missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Acetate, Crystal, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1015500GM
Specifications
| 5743-04-4 | |
| Poly Bottle | |
| 500 g | |
| O.O.[Cd++].CC([O-])=O.CC([O-])=O | |
| 266.53 | |
| Reagent |
| 1 | |
| C4H10CdO6 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| cadmium(2+) diacetate dihydrate | |
| 0.98 | |
| 0.0001 |
Chemical Identifiers
| 5743-04-4 | |
| 266.53 | |
| cadmium(2+) diacetate dihydrate |
| C4H10CdO6 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| O.O.[Cd++].CC([O-])=O.CC([O-])=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.