missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate, Crystal, 98%, Spectrum™ Chemical

Catalog No. 18609389
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609389 25 g Poly Bottle
18609390 125 g Poly Bottle
18609392 500 g Poly Bottle
18609393 2.5 kg Poly Pail
18609391 12 kg Poly Pail
18609394 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609389 Supplier Spectrum Chemical Mfg Cor Supplier No. C101525GM

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L
IUPAC Name cadmium(2+) diacetate dihydrate
SMILES O.O.[Cd++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 25 g
Percent Purity 0.98
Grade Reagent
Insoluble Matter 0.0001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.