missing translation for 'onlineSavingsMsg'
Learn More

Cadmium, Coarse powder, 0.3-1.6 mm, EMSURE™ MilliporeSigma™

Catalog No. M1020010250
Change view
Click to view available options
Quantity:
250 g
Packaging:
Metal can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1020010250 250 g Metal can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1020010250 Supplier Technidata Supplier No. 1.02001.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coarse powder. Used for analysis and for filling reductors.

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Melting Point 321°C
Density 8.64 g/cm3 (20 °C)
Quantity 250 g
Packaging Metal can
UN Number UN 3288
Formula Weight 112.41
Hazard Category Very toxic
Dangerous for the environment
Hazard Statement R 45-26-48/23/25-68-50/53-62-63
May cause cancer.Also very toxic by inhalation.Also toxic: danger of serious damage to health by prolonged exposure through inhalation and if swallowed.Possible risk of irreversible effects.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.Possible risk of impaired fertility.Possible risk of harm to the unborn child.
Precautionary Statement S 53-45-60-61
Avoid exposure - obtain special instructions before use.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).This material and its container must be disposed of as hazardous waste.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
DOTInformation 6.1, III
EINECSNumber 231-152-8
storageNote1 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
Recommended Storage Store below +30°C.