missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadence Science™ Reusable Animal Feeding Needles: Newborn

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

Supplier:  Cadence Science™ POP7900

 View more versions of this product

Catalog No. 01-290-1



Description

Description

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

For newborns.

  • For the intubation of laboratory animals
  • Straight, curved tubing styles
  • Many standard sizes available, from small to large
  • Straight
  • Autoclavable

Newborn Needles

  • For newborn mice and rats
  • For animals up to 0.7 oz. (20g)

Specifications

Specifications

Autoclavable
Straight Animal Feeding Needle
Newborn Mice and Rat, ideal for feeding and dosing
1 in.
0.049 in.
Stainless Steel
No
24G
0.254 cm
0.013 cm
SDS
Documents

Documents

Promotions

Promotions