Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Supplier:  Cadence Science™ POP9927
 View more versions of this product
Catalog No. 14825259
Only null left
Description
Description
  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box
Specifications
Chemical Identifiers
Soft, Flexible Animal Feeding Needle
PTFE
Single-use
Sterile
Specifications
Soft, Flexible Animal Feeding Needle
Single-use
15G
3.9 in.
0.11 in.
PTFE
For critical applications requiring sterile, flexible, single use feeding needles
Sterile
1 cm
0.028 cm
Product Suggestions
Product Suggestions