missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
Description
- Nontoxic, nonpyrogenic
- PTFE tubing is flexible and softer than other types of tubing
- Low friction protects delicate organs
- Translucent for easy confirmation of reagent delivery
- Plastic luer hubs; soft silicone tips
- Sterile, packaged in pouches
- Five needles per pouch, 20 pouches per box
Specifications
Specifications
| Product Type | Soft, Flexible Animal Feeding Needle |
| Material | PTFE |
| Disposable | Single-use |
| For Use With (Application) | For critical applications requiring sterile, flexible, single use feeding needles |
| Gauge | 18G |
| Sterility | Sterile |
| Length (English) | 3 in. |
| Length (Metric) | 0.75 cm |
| Diameter (English) Ball | 0.078 in. |
| Diameter (Metric) Ball | 0.02 cm |