missing translation for 'onlineSavingsMsg'
Learn More

Cabazitaxel, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 468895000

 View more versions of this product

Catalog No. AC468895000


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cabazitaxel
Available
Powder
MFCD18827611
170°C to 180°C
COC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C4=CC=CC=C4)C(C)=C(C(OC)C(=O)C12C)C3(C)C
835.94
Glass Bottle
183133-96-2
C45H57NO14
White to Yellow to Beige
BMQGVNUXMIRLCK-UHFFFAOYNA-N
4-(acetyloxy)-15-[(3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-3-phenylpropanoyl)oxy]-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
500 mg

Chemical Identifiers

183133-96-2
835.94
BMQGVNUXMIRLCK-UHFFFAOYNA-N
COC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C4=CC=CC=C4)C(C)=C(C(OC)C(=O)C12C)C3(C)C
C45H57NO14
MFCD18827611
4-(acetyloxy)-15-[(3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-3-phenylpropanoyl)oxy]-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Health hazard

Exclamation mark

Recommended Storage : Normal conditions

SDS
Documents

Documents

Promotions

Promotions