missing translation for 'onlineSavingsMsg'
Learn More

Cabazitaxel, Thermo Scientific™

Catalog No. AC468891000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468891000 100 mg
AC468895000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468891000 Supplier Thermo Scientific Chemicals Supplier No. 468891000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 183133-96-2
Molecular Formula C45H57NO14
Molecular Weight (g/mol) 835.94
MDL Number MFCD18827611
InChI Key BMQGVNUXMIRLCK-UHFFFAOYNA-N
IUPAC Name 4-(acetyloxy)-15-[(3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-3-phenylpropanoyl)oxy]-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES COC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C4=CC=CC=C4)C(C)=C(C(OC)C(=O)C12C)C3(C)C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Yellow to Beige
Melting Point 170°C to 180°C
Quantity 100 mg
Chemical Name or Material Cabazitaxel
Hazard Category Exclamation mark
Hazard Statement Health hazard
Recommended Storage Normal conditions