missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150283
Description
CA7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known; however, it may have a role in inherited renal abnormalities of bicarbonate transport.Chemical Identifiers
| CA7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P43166, Q9ERQ8 | |
| 12354, 291819, 766 | |
| Recombinant Carbonic Anhydrase VII (CA7) (Ser58-Pro217). | |
| Antibody |
| 0.5 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Ca7 | |
| AV343731; CA7; Car7; Carbonate dehydratase VII; carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII; CAVII; CA-VII | |
| Ca7, Car7 | |
| Primary | |
| Protein A |
Specifications
| CA7 | |
| Polyclonal | |
| Unconjugated | |
| Ca7 | |
| AV343731; CA7; Car7; Carbonate dehydratase VII; carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII; CAVII; CA-VII | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12354, 291819, 766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P43166, Q9ERQ8 | |
| Ca7, Car7 | |
| Recombinant Carbonic Anhydrase VII (CA7) (Ser58-Pro217). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |