missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf89 Monoclonal Antibody (OTI4E10)

Description
C9orf89 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | C9orf89 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4E10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CARD19 |
| Gene Accession No. | Q96LW7 |
| Gene Alias | 1110007C09Rik; AI851695; Bcl10-interacting CARD protein; Bcl10-interacting protein with CARD; BinCARD; C9orf89; CARD19; caspase recruitment domain family member 19; caspase recruitment domain family, member 19; caspase recruitment domain-containing protein 19; RIKEN cDNA 1110007C09 gene |
| Show More |