missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C9orf72 Recombinant Rabbit Monoclonal Antibody (PSH11-11)
GREENER_CHOICE

Catalog No. MA569959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA569959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA569959 Supplier Invitrogen™ Supplier No. MA569959

Rabbit Recombinant Monoclonal Antibody

Positive control: SH-SY5Y cell lysate, Neuro-2a cell lysate, Human brain tissue lysate, Mouse brain tissue lysate, Rat brain tissue lysate. Subcellular location: Nucleus, Cytoplasm.

C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.

Specifications

Antigen C9orf72
Applications Western Blot
Classification Recombinant Monoclonal
Clone PSH11-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide, pH 7.4
Gene C9orf72
Gene Accession No. Q66HC3, Q96LT7
Gene Alias 3110043O21Rik, AI840585, ALSFTD, C9orf72, chromosome 9 open reading frame 72, FTDALS, FTDALS1, Guanine nucleotide exchange C9orf72, Guanine nucleotide exchange C9orf72 homolog, protein C9orf72, protein C9orf72 homolog, RGD1359108, RIKEN cDNA 3110043O21 gene, similar to RIKEN cDNA 3110043O21
Gene Symbols C9orf72, RGD1359108
Host Species Rabbit
Immunogen Recombinant protein within human C9orf72 aa 1-250.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203228, 313155, 73205
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less