missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C4BPB Recombinant Rabbit Monoclonal Antibody (23GB3715)

Catalog No. MA552629
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552629 20 μL
MA552628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552629 Supplier Invitrogen™ Supplier No. MA552629
Only null left

Rabbit Recombinant Monoclonal Antibody

C4BPB Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Complement component 4-binding protein (C4BP) is a plasma glycoprotein that inhibits the classical pathway of complement activation, which is mediated through antibody targeting of foreign antigen. Structurally, C4BP is a disulfide linked, multimeric protein that is composed of seven alpha chains and one beta chain. C4BP functions as a cofactor for C3beta inactivator in the cleavage of C3beta, and accelerates the decay of C4betaC2alpha (C3 convertase) by acting as a cofactor in the cleavage of C4beta by factor I. Streptococcal strains that express Ig-binding cell surface molecules, which are members of the M protein family, can bind to overlapping C4beta binding sites in C4BP and therefore, interfere with the classical pathway of complement activation. Bacteria-bound C4BP may be an evolved mechanism that downregulates complement activation in the bacterial host microenvironment, thereby reducing the occurrences of bacterial opsonization and phagocytosis.

Specifications

Antigen C4BPB
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3715
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C4BPB
Gene Accession No. P20851
Gene Alias C4b-binding protein beta chain; C4BP; C4BPB; C4bp-ps1; complement component 4 binding protein beta; complement component 4 binding protein, beta; complement component 4 binding protein, pseudogene 1
Gene Symbols C4BPB
Host Species Rabbit
Immunogen A synthetic peptide of human C4 binding protein (1-252AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 725
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less