missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4 - C24 Even Carbon Saturated FAMEs, 1,000 μg/mL Each Component in Hexane, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Physical Form | Liquid |
| Concentration | 1000 μg/mL each component in hexane |
| Quantity | 1 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Methyl arachidate; Methyl behenate; Methyl butyrate; Methyl decanoate; Methyl hexanoate; Methyl dodecanoate; Methyl myristate; Methyl octanoate; Methyl palmitate; Methyl stearate; Methyl tetracosanoate |