missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, Trihydrate, Granular, Technical, Spectrum™ Chemical

Catalog No. 18612628
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612628 50 kg Amber Glass Bottle
18612625 500 g Amber Glass Bottle
18612626 2.5 kg Amber Glass Bottle
18612627 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612628 Supplier Spectrum Chemical Mfg Cor Supplier No. P129650KGBL
Get Up to $300 in Additional Chemicals
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14459-95-1

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 50 kg
Molecular Weight (g/mol) 422.39
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Ferrocyanide, Trihydrate, Granular

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.