missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3orf38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121394
Description
Antibody tested in Indirect ELISA.
Chemical Identifiers
| C3orf38 | |
| -20°C or -80°C if preferred | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| C3orf38 | |
| C3orf38; chromosome 3 open reading frame 38; uncharacterized protein C3orf38 | |
| C3orf38 | |
| Primary | |
| Protein G |
| 4.2 mg/mL | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5JPI3 | |
| 285237 | |
| Recombinant Human Uncharacterized protein C3orf38 protein (80-329aa). | |
| Antibody |
Specifications
| C3orf38 | |
| Polyclonal | |
| Unconjugated | |
| C3orf38 | |
| C3orf38; chromosome 3 open reading frame 38; uncharacterized protein C3orf38 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 285237 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 4.2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5JPI3 | |
| C3orf38 | |
| Recombinant Human Uncharacterized protein C3orf38 protein (80-329aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |