missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C20orf177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559397 Supplier Invitrogen™ Supplier No. PA559397

Rabbit Polyclonal Antibody

Immunogen sequence: SSKSTKLQRW DLSGSGSSSK VETSGHIRVP KQAAVILDSA DSCKASKTQA HAHPRKKGKA ESCGHATVSS EKKLKTNGVK Q Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 43%.

FAM217B is a protein coding gene.

Specifications

Antigen C20orf177
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM217B
Gene Accession No. Q9NTX9
Gene Alias C20orf177; dJ551D2.5; FAM217B; family with sequence similarity 217 member B; family with sequence similarity 217, member B; protein FAM217B; uncharacterized protein C20orf177
Gene Symbols FAM217B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C20orf177. Recombinant protein control fragment (Product #RP-99461).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less