missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QBP Recombinant Rabbit Monoclonal Antibody (23GB695)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | C1QBP |
|---|---|
| Clone | 23GB695 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
C1QBP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.Specifications
| C1QBP | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| C1QBP | |
| AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
| C1QBP | |
| Primary | |
| Affinity chromatography |
| 23GB695 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07021 | |
| 708 | |
| A synthetic peptide of human GC1q R (150-200AA). | |
| Antibody |