missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QBP Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5149000
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.Chemical Identifiers
| C1QBP | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human, Mouse, Rat | |
| C1QBP | |
| AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
| C1QBP | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with trehalose and no preservative | |
| O35658, O35796, Q07021 | |
| 12261, 29681, 708 | |
| E. coli-derived recombinant mouse HABP1/C1QBP , Leu72-Gln279. | |
| Antibody |
Specifications
| C1QBP | |
| Polyclonal | |
| Unconjugated | |
| C1QBP | |
| AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 12261, 29681, 708 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O35658, O35796, Q07021 | |
| C1QBP | |
| E. coli-derived recombinant mouse HABP1/C1QBP , Leu72-Gln279. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |