missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf226 Polyclonal Antibody

Description
Immunogen sequence: DFLRRKEPSS LGSVGVTEIN KTAGAQLASG TDAAPEAWLE DERSVLQETF PRLDPPPPIT RKRTPRALKT TQD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
C1orf226 (Chromosome 1 Open Reading Frame 226) is a Protein Coding gene. Diseases associated with C1orf226 include Phosphoglycerate Dehydrogenase Deficiency. An important paralog of this gene is DAB2.
Specifications
Specifications
| Antigen | C1orf226 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C1orf226 |
| Gene Accession No. | A1L170 |
| Gene Alias | C1orf226; chromosome 1 open reading frame 226; uncharacterized protein C1orf226 |
| Gene Symbols | C1orf226 |
| Show More |