missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C17orf49 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531990
Description
Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Component of chromatin complexes such as the MLL1/MLL and NURF complexes.Chemical Identifiers
| C17orf49 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| C17orf49 | |
| 0610010K14Rik; 1110020A23Rik; AL033328; AU045833; BAP18; BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; C17orf49; Chromatin complexes subunit BAP18; chromosome 17 open reading frame 49; HEPIS; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49; MLL1/MLL complex subunit C17orf49 homolog; RIKEN cDNA 0610010K14 gene | |
| C17orf49 | |
| Primary | |
| Antigen affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IXM2 | |
| 124944 | |
| Recombinant protein encompassing a sequence within the center region of human C17orf49. The exact sequence is proprietary. | |
| Antibody |
Specifications
| C17orf49 | |
| Polyclonal | |
| Unconjugated | |
| C17orf49 | |
| 0610010K14Rik; 1110020A23Rik; AL033328; AU045833; BAP18; BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; C17orf49; Chromatin complexes subunit BAP18; chromosome 17 open reading frame 49; HEPIS; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49; MLL1/MLL complex subunit C17orf49 homolog; RIKEN cDNA 0610010K14 gene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124944 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IXM2 | |
| C17orf49 | |
| Recombinant protein encompassing a sequence within the center region of human C17orf49. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |