missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals C16 Ceramide, 98%

Catalog No. p-7070114
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67093MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67093MCR Supplier Thermo Scientific Chemicals Supplier No. J67093MCR
Only null left

An endogenous ceramide that acts as a lipid second messenger to regulate apoptosis and stress signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24696-26-2
Molecular Formula C34H67NO3
Synonym D-erythro-N-Palmitoylsphingosine; N-Hexadecanoyl-D-erythro-sphingosine

Specifications

Quantity 1 mg
Content And Storage -30°C to -10°C
For Use With (Application) An endogenous ceramide that acts as a lipid second messenger to regulate apoptosis and stress signaling.
Formula Weight 537.91
Solubility Soluble in DMF at 0.15mg/ml
Product Type C16 Ceramide, 98%
Form Crystalline solid
InChI Key YDNKGFDKKRUKPY-UHFFFAOYNA-N
SMILES CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
IUPAC Name N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Molecular Weight (g/mol) 537.91
Assay 98%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only