CAS RN 24696-26-2
IUPAC Name:
N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Synonyms:
N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Molecular Weight (g/mol):
537.91
Molecular Formula:
C34H67NO3
InChi Key:
YDNKGFDKKRUKPY-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
1
–
2
of
2
results
1
Thermo Scientific Chemicals C16 Ceramide, 98%
CAS: 24696-26-2 Molecular Formula: C34H67NO3 Synonym: D-erythro-N-Palmitoylsphingosine; N-Hexadecanoyl-D-erythro-sphingosine
2
Promotions Available
N-Palmitoyl-D-erythro-sphingosine, synthetical, 98%
N-Palmitoyl-D-erythro-sphingosine, synthetical, 98%
CAS: 24696-26-2 Molecular Formula: C34H67NO3 Molecular Weight (g/mol): 537.91 MDL Number: MFCD00056205 InChI Key: YDNKGFDKKRUKPY-UHFFFAOYNA-N Synonym: c16-ceramide,d-erythro-c16-ceramide,n-palmitoyl 4-sphingenine,n-palmitoyl-d-sphingosine,d-erythro-4-ceramide,n-palmitoylsphingosinee,d-erythro-n-palmitoylsphingosine,n-hexadecanoyl-d-erythro-ceramide,d-erythro-n-hexadecanoylsphingenine,2s,3r,4e-2-n-palmitoyloctadecasphinga-4-ene PubChem CID: 71314645 SMILES: CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC