missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12orf32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144036
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144036 Supplier Invitrogen™ Supplier No. PA5144036
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MOLT-4 whole cell, human U251 whole cell. Flow: RT4 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

RHNO1 plays a role in DNA damage response (DDR) signaling upon genotoxic stresses such as ionizing radiation (IR) during the S phase. Recruited to sites of DNA damage through interaction with the 9-1-1 cell-cycle checkpoint response complex and TOPBP1 in a ATR-dependent manner. Required for the progression of the G1 to S phase transition. Plays a role in the stimulation of CHEK1 phosphorylation.

Specifications

Antigen C12orf32
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene RHNO1
Gene Accession No. Q9BSD3
Gene Alias C12orf32; HKMT1188; RAD9, HUS1, RAD1-interacting nuclear orphan protein 1; Rad9, Rad1, Hus1 interacting nuclear orphan; rAD9, RAD1, HUS1-interacting nuclear orphan protein; RAD9-HUS1-RAD1 interacting nuclear orphan 1; RHINO; RHNO1
Gene Symbols RHNO1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human RHNO1 (RQPSQKAPLLFHQQPLE) (aa 8-24).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83695
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less