missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C11orf9 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Zebrafish: 79%, Rat: 100%.
MYRF is a protein coding gene.
Specifications
Specifications
| Antigen | C11orf9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | MYRF |
| Gene Accession No. | Q9Y2G1 |
| Gene Alias | 6030439E18; C11orf9; C18H11orf9; Gm1804; Gm98; KIAA0954; Mrf; myelin gene regulatory factor; myelin gene regulatory factor; myelin regulatory factor; myelin gene regulatory factor-like; myelin regulatory factor; Myelin regulatory factor, C-terminal; Myelin regulatory factor, N-terminal; MYRF; Ndt80; pqn-47; RGD1306622; zgc:172183 |
| Gene Symbols | MYRF |
| Show More |