missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C10orf113 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557915
Description
Immunogen sequence: LQSWPTLARW LRRVSFSLYK PPIQAAPEPD PGNWAKSPRL SLGPEGITKG KHGFKFQGIK EKFNVSKKVL K Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 32%.
Nebulette encodes a nebulin like protein that is abundantly expressed in cardiac muscle. The encoded protein binds actin and interacts with thin filaments and Z-line associated proteins in striated muscle. This protein may be involved in cardiac myofibril assembly. A shorter isoform of this protein termed LIM nebulette is expressed in non-muscle cells and may function as a component of focal adhesion complexes. Alternate splicing results in multiple transcript variants.Specifications
| C10orf113 | |
| Polyclonal | |
| Unconjugated | |
| NEBL | |
| 1200007O21Rik; A630080F05Rik; Actin-binding Z-disk protein; BB140644; D830029A09Rik; LASP2; LIM and SH3 protein 2; LIM zinc-binding domain-containing Nebulette; LIM-nebulette; Lnebl; Nebl; Nebulette | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10529 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76041 | |
| NEBL | |
| Recombinant protein corresponding to Human C10orf113. Recombinant protein control fragment (Product #RP-97225). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |