missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-MAF Monoclonal Antibody (sym0F1), PE-eFluor™ 610, eBioscience™

Catalog No. 61985542
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61985542 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61985542 Supplier Invitrogen™ Supplier No. 61985542
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

The sym0F1 monoclonal antibody reacts with human and mouse c-Maf, a 42 kDa basic leucine zipper transcription factor shown to be involved in the neural, ocular and hematopoietic systems. In hematopoietic cells, it was first shown to be crucial for IL-4 expression in Th2 and was the first transcription factor believed to be Th subset-specific. More recent evidence shows that specific phospho-tyrosine residues lead to upregulation of IL-4. c-Maf has also been shown to be important to differentiation and function in both Th17 and Tfh cells. It drives expression of IL-21 in both cell types, while promoting expression of IL-23R in Th17 and CXCR5 in Tfh as well. Sym0F1 does not cross-react with MafA or MafB. Applications Reported: This sym0F1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This sym0F1 antibody has been pre-diluted and tested by intracellular staining followed by flow cytometric analysis of Th17-polarized mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to 'Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins' located at Flow Protocols . This may be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirical...

c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.

Specifications

Antigen c-MAF
Applications Flow Cytometry
Classification Monoclonal
Clone sym0F1
Concentration 5 μL/Test
Conjugate PE-eFluor 610
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene MAF
Gene Accession No. O75444, P54843
Gene Alias 2810401A20Rik; A230108G15Rik; Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene; avian musculoaponeurotic fibrosarcoma (v-maf) AS42 oncogene homolog; AW047063; AYGRP; basic domain leucine zipper transcription factor; CCA4; cMaf; C-Maf; c-Maf long form; c-maf proto-oncogene; CTRCT21; fl13d12; hMafA; LOW QUALITY PROTEIN: transcription factor Maf; LOW QUALITY PROTEIN: transcription factor MafA; maf; MAF bZIP transcription factor; MAF bZIP transcription factor A; maf protein; Maf2; MAFA; Mafa homolog; MGC71685; pancreatic beta-cell-specific transcriptional activator; Proto-oncogene c-Maf; RGD1562627; RIPE3b1; RIPE3b1 factor; T lymphocyte c-maf long form; transcription factor Maf; Transcription factor Maf-2; transcription factor MafA; transcription factor RIPE3b1; v-maf avian musculoaponeurotic fibrosarcoma oncogene A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog a (paralog a); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog; V-maf musculoaponeurotic fibrosarcoma oncogene homolog A; wu:fl13d12; Z-cmaf
Gene Symbols MAF
Host Species Mouse
Immunogen Full-length E. coli-produced c-Maf.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17132, 4094
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less