missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyryl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 203805-93-0 |
| Linear Formula | C11D3H18 NO4 |
| MDL Number | MFCD01073487 |
| Synonym | (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxobutoxy)-1-propanaminium inner salt; Butanoyl-(methyl-d3)-L-carnitine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 234.31 |
| Percent Purity | ≥97.0% (TLC) |
| Grade | Analytical Standard |