missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyrophenone 99.0+%, TCI America™
$73.16 - $685.36
Chemical Identifiers
| CAS | 495-40-9 |
|---|---|
| Molecular Formula | C10H12O |
| Molecular Weight (g/mol) | 148.205 |
| MDL Number | MFCD00009397 |
| InChI Key | FFSAXUULYPJSKH-UHFFFAOYSA-N |
| Synonym | butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone |
| PubChem CID | 10315 |
| IUPAC Name | 1-phenylbutan-1-one |
| SMILES | CCCC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B076925ML
|
TCI America
B076925ML |
25 mL |
Each for $73.16
|
|
|||||
|
B0769100ML
|
TCI America
B0769100ML |
100 mL |
Each for $223.33
|
|
|||||
|
B0769500ML
|
TCI America
B0769500ML |
500 mL |
Each for $685.36
|
|
|||||
Chemical Identifiers
| 495-40-9 | |
| 148.205 | |
| FFSAXUULYPJSKH-UHFFFAOYSA-N | |
| 10315 | |
| CCCC(=O)C1=CC=CC=C1 |
| C10H12O | |
| MFCD00009397 | |
| butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone | |
| 1-phenylbutan-1-one |
Specifications
| 495-40-9 | |
| Yellow | |
| C10H12O | |
| 25 mL | |
| FFSAXUULYPJSKH-UHFFFAOYSA-N | |
| 1-phenylbutan-1-one | |
| 10315 | |
| ≥99.0% (GC) | |
| Butyrophenone |
| 12°C | |
| 221°C | |
| MFCD00009397 | |
| butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone | |
| CCCC(=O)C1=CC=CC=C1 | |
| 148.205 | |
| 148.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2673
TSCA : Yes