missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone 99.0+%, TCI America™

Catalog No. B076925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B076925ML 25 mL
B0769100ML 100 mL
B0769500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B076925ML Supplier TCI America Supplier No. B076925ML

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009397
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 12°C
Color Yellow
Boiling Point 221°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyrophenone
EINECSNumber (3)-2673
TSCA Yes