missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, 99%

CAS: 495-40-9 | C10H12O | 148.205 g/mol

$65.67 - $451.92

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009397
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1134514
View Documents
Thermo Scientific Chemicals
A1134514
25 g
Each for $65.67
Only null left
 
AAA1134518
View Documents
Thermo Scientific Chemicals
A1134518
50 g
Each for $126.38
Only null left
 
AAA1134530
View Documents
Thermo Scientific Chemicals
A1134530
250 g
Each for $451.92
Only null left
 
Description

Description

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
Specifications

Chemical Identifiers

495-40-9
148.205
FFSAXUULYPJSKH-UHFFFAOYSA-N
10315
CCCC(=O)C1=CC=CC=C1
C10H12O
MFCD00009397
butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
1-phenylbutan-1-one

Specifications

495-40-9
1.021
88°C (190°F)
C10H12O
MFCD00009397
508305
Insoluble in water.
CCCC(=O)C1=CC=CC=C1
148.205
148.21
Butyrophenone
11°C to 13°C
222°C
Characteristic
1.5195
25 g
butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
FFSAXUULYPJSKH-UHFFFAOYSA-N
1-phenylbutan-1-one
10315
99%
Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P370+P378q-P374-P501c

H227

EINECSNumber : 207-799-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only