missing translation for 'onlineSavingsMsg'
Learn More

Butyronitrile 99.0+%, TCI America™

Catalog No. B076825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B076825ML 25 mL
B0768500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B076825ML Supplier TCI America Supplier No. B076825ML

Chemical Identifiers

CAS 109-74-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Specifications

Color Colorless
Boiling Point 118°C
Quantity 25 mL
UN Number 2411
Formula Weight 69.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyronitrile
EINECSNumber (2)-3910&(2)-3924
RTECSNumber ET8750000
TSCA Yes