Learn More
Butyramide, 98%
CAS: 541-35-5 | C4H9NO | 87.122 g/mol
Supplier: Thermo Scientific Chemicals A1249422
Description
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsButyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.
Solubility
Soluble in water (170 g/L).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Butyramide | |
| 113°C to 116°C | |
| 216°C | |
| MFCD00041894 | |
| 1361528 | |
| butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid | |
| DNSISZSEWVHGLH-UHFFFAOYSA-N | |
| butanamide | |
| 10927 | |
| 87.12 |
| 541-35-5 | |
| 1.032 | |
| C4H9NO | |
| 100 g | |
| 14,1592 | |
| Soluble in water (170g/L). | |
| CCCC(=O)N | |
| 87.122 | |
| CHEBI:50724 | |
| 98% |
Chemical Identifiers
| 541-35-5 | |
| 87.122 | |
| DNSISZSEWVHGLH-UHFFFAOYSA-N | |
| 10927 | |
| butanamide |
| C4H9NO | |
| MFCD00041894 | |
| butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid | |
| CHEBI:50724 | |
| CCCC(=O)N |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 208-776-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only