missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™

Catalog No. p-7119140
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15941G 1 g
B159410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15941G Supplier TCI America Supplier No. B15941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1527-98-6
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00191327
InChI Key IKGRHEWIFBFXPP-IZZDOVSWSA-N
Synonym butanal, 2,4-dinitrophenyl hydrazone, butyraldehyde-2,4-dinitrophenylhydrazone, n-butyraldehyde 2,4-dinitrophenylhydrazone, butyraldehyde, 2,4-dinitrophenyl hydrazone, 1e-1-azapent-1-enyl 2,4-dinitrophenyl amine, 1-butylidene-2-2,4-dinitrophenyl hydrazine, 1e-butanal 2,4-dinitrophenyl hydrazone #, butyraldehyde-2,4-dnph, butanal,4-dinitrophenyl hydrazone, butyraldehyde,4-dinitrophenyl hydrazone
PubChem CID 9601272
IUPAC Name (1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
SMILES CCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 122°C
Quantity 1 g
UN Number 1325
Formula Weight 252.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyraldehyde 2,4-Dinitrophenylhydrazone