accessibility menu, dialog, popup

UserName

H3CNNHNOONOO(E)

CAS RN 1527-98-6

IUPAC Name: (1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
Synonyms: (1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
Molecular Weight (g/mol): 252.23
Molecular Formula: C10H12N4O4
InChi Key: IKGRHEWIFBFXPP-IZZDOVSWSA-N
SMILES: CCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)