missing translation for 'onlineSavingsMsg'
Learn More

Butyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. B097025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B097025G 25 g
B0970250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B097025G Supplier TCI America Supplier No. B097025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1779-51-7
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00011855
InChI Key IKWKJIWDLVYZIY-UHFFFAOYSA-M
Synonym butyltriphenylphosphonium bromide, n-butyltriphenylphosphonium bromide, 1-butyl triphenylphosphonium bromide, phosphonium, butyltriphenyl-, bromide, butyl triphenyl phosphonium bromide, butyl triphenyl phosphine bromide, n-butyltriphenylphosphonium chloride, butyltriphenylphosphanium bromide, phosphonium, butyltriphenyl-, bromide 1:1, n-butyl triphenylphosphonium bromide
PubChem CID 159628
IUPAC Name butyltriphenylphosphanium bromide
SMILES [Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 243°C
Color White
Quantity 25 g
UN Number 3464
Formula Weight 399.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyltriphenylphosphonium Bromide
EINECSNumber (3)-4338
RTECSNumber TA1855200
TSCA Yes