missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 94-26-8 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Physical Form | Neat |
| Linear Formula | C11H14O3 |
| MDL Number | MFCD00016478 |
| Synonym | 4-Hydroxybenzoic acid butyl ester |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 194.23 |
| Grade | Certified Reference Material |