missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylparaben, NF, 98-102%, Spectrum™ Chemical
BU115, 94-26-8, C11H14O3
Supplier: Spectrum Chemical Mfg Cor BU11525KG
Description
Spectrum™ Chemical Butylparaben, NF is an antimicrobial preservative used as a suspending agent for medications. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 94-26-8 | |
| 0.1% | |
| C11H14O3 | |
| QFOHBWFCKVYLES-UHFFFAOYSA-N | |
| butyl 4-hydroxybenzoate | |
| 98-102% | |
| Crystalline Powder or Lumps |
| 100% | |
| Poly Bottle | |
| 2.5 kg | |
| CCCCOC(=O)C1=CC=C(O)C=C1 | |
| 194.23 | |
| NF |
Chemical Identifiers
| 94-26-8 | |
| 194.23 | |
| butyl 4-hydroxybenzoate |
| C11H14O3 | |
| QFOHBWFCKVYLES-UHFFFAOYSA-N | |
| CCCCOC(=O)C1=CC=C(O)C=C1 |