missing translation for 'onlineSavingsMsg'
Learn More

Butylparaben, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18601171
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601171 125 g Poly Bottle
18601172 500 g Poly Bottle
18601173 2.5 kg Poly Bottle
18601174 12 kg Poly Bottle
18601175 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601171 Supplier Spectrum Chemical Mfg Cor Supplier No. BU115125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

BU115, 94-26-8, C11H14O3

Spectrum™ Chemical Butylparaben, NF is an antimicrobial preservative used as a suspending agent for medications. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=C(O)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 98 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps