missing translation for 'onlineSavingsMsg'
Learn More

Butylone hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. B0451ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0451ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0451ML Supplier MilliporeSigma Supplier No. B0451ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Butylone, also known as bk-MBDB or B1, is a designer drug and stimulant with cases of abuse reported in the US and Europe.
  • This certified solution standard is suitable for numerous GC/MS and LC/MS applications including clinical toxicology testing, forensic analysis, urine drug testing or calibrator preparation.

Specifications

Chemical Name or Material Butylone HCl
Packaging Ampule
CAS 17762-90-2
Molecular Formula C12H16ClNO3
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 257.71
Quantity 1 mL
Grade Certified
Show More Show Less