missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxytoluene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019624
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019624 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019624 Supplier Merck Emd Millipore Supplier No. PHR11171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Butylated hydroxytoluene (BHT) is a phenolic antioxidant used as a food additive. It is frequently added to dried cereals, cooking oils and animal foods. BHT is also known to enhance the stability of pharmaceuticals, fat-soluble vitamins and cosmetics . Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 128-37-0
Ignition Point 878°F
Vapor Pressure <0.01 mmHg (20°C)
Vapor Density 7.6 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula [(CH3)3 C]2C6H2(CH3)OH
MDL Number MFCD00011644
Melting Point 69°C to 73°C
Boiling Point 265°C
Synonym 2,6-Di-tert-butyl-4-methylphenol; 2,6-Di-tert-butyl-p-cresol; BHT; Butylated hydroxytoluene; Butylhydroxytoluene; DBPC
Type Certified Reference Material
Quantity 1 g
Formula Weight 220.35
Grade Certified Reference Material
Show More Show Less