missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxytoluene, 99.5%, MP Biomedicals™

Catalog No. p-4404317
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10116290 500 g
ICN10116291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10116290 Supplier MP Biomedicals Inc Supplier No. 0210116290

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Density 1.048g/mL at 20°C
Assay Percent Range 99.5%
Quantity 500 g
Formula Weight 220.36
Physical Form Crystalline
Chemical Name or Material Butylated hydroxytoluene