missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxyanisole, 98.5%, MP Biomedicals™

Catalog No. ICN10115990
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10115990 500 g
ICN10115980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10115990 Supplier MP Biomedicals Inc Supplier No. 0210115990

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Assay Percent Range 98.5%
Quantity 500 g
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material Butylated Hydroxyanisole
Recommended Storage Store at room temperature (15° to 30°C). Protect from light.