Learn More
Butylated hydroxyanisole, 96%
CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol
Supplier: Thermo Scientific Chemicals 235230025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Butylated hydroxyanisole | |
| 25013-16-5 | |
| White to Yellow | |
| 113°C | |
| 95% min. (GC) | |
| C11H16O2 | |
| 2.5 kg | |
| 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
| MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
| 2-tert-butyl-4-methoxyphenol | |
| 8456 | |
| 180.24 | |
| Crystalline Flakes or Crystals |
| 96% | |
| 48°C to 55°C | |
| 268°C | |
| Authentic | |
| Plastic bottle | |
| MFCD01779059 | |
| 15,1549 | |
| Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,propylene glycol,,chlorogorm,ether and acetone,slightly soluble in benzene | |
| CC(C)(C)C1=C(C=CC(=C1)OC)O | |
| 180.24 | |
| CHEBI:76358 | |
| 96% |
Chemical Identifiers
| 25013-16-5 | |
| 180.24 | |
| MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
| 8456 | |
| 2-tert-butyl-4-methoxyphenol |
| C11H16O2 | |
| MFCD01779059 | |
| 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
| CHEBI:76358 | |
| CC(C)(C)C1=C(C=CC(=C1)OC)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED:
GHS Signal Word: Warning
EINECSNumber : 246-563-8
RTECSNumber : SL1945000
TSCA : TSCA
RUO – Research Use Only