missing translation for 'onlineSavingsMsg'
Learn More

Butylamine Hydroiodide 97.0+%, TCI America™

Catalog No. B44335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44335G 5 g
B44331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44335G Supplier TCI America Supplier No. B44335G
Only null left

Chemical Identifiers

CAS 36945-08-1
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.051
InChI Key CALQKRVFTWDYDG-UHFFFAOYSA-N
Synonym Butylammonium Iodide
PubChem CID 88075134
IUPAC Name butan-1-amine;hydroiodide
SMILES CCCCN.I

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.05
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Butylamine Hydroiodide