accessibility menu, dialog, popup

UserName

CAS RN 36945-08-1

CH3H2NHI

CAS RN 36945-08-1

IUPAC Name: butan-1-amine hydroiodide
Synonyms: butylamine hydroiodide
Molecular Weight (g/mol): 201.05
Molecular Formula: C4H12IN
InChi Key: CALQKRVFTWDYDG-UHFFFAOYSA-N
SMILES: I.CCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results