missing translation for 'onlineSavingsMsg'
Learn More

Butylamine, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019599
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019599 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019599 Supplier Merck Emd Millipore Supplier No. 908931ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 109-73-9
Ignition Point 594°F
Vapor Pressure 68 mmHg (20°C)
Vapor Density 2.5 (vs. air)
Density 0.74 g/mL (at 25°C)
Flash Point -7°C
Physical Form Neat
Linear Formula CH3(CH2)3 NH2
MDL Number MFCD00011690
Melting Point -49°C
Boiling Point 78°C
Synonym 1-Aminobutane; n-Butylamine
Type Analytical Standard
Quantity 1 mL
Formula Weight 73.14
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less