missing translation for 'onlineSavingsMsg'
Learn More

Butyl Vinyl Ether (stabilized with KOH) 98.0+%, TCI America™

Catalog No. B074625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074625ML 25 mL
B0746500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B074625ML Supplier TCI America Supplier No. B074625ML

Chemical Identifiers

CAS 111-34-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009454
InChI Key UZKWTJUDCOPSNM-UHFFFAOYSA-N
Synonym n-butyl vinyl ether, butyl vinyl ether, vinyl butyl ether, butane, 1-ethenyloxy, butoxyethylene, 1-ethenyloxy butane, butoxyethene, ether, butyl vinyl, vinyl n-butyl ether, ethenyl n-butyl ether
PubChem CID 8108
IUPAC Name 1-(ethenyloxy)butane
SMILES CCCCOC=C

Specifications

Melting Point -92°C
Color Yellow
Boiling Point 93°C
Quantity 25 mL
UN Number 2352
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Vinyl Ether (stabilized with KOH)
EINECSNumber (2)-0372
RTECSNumber KN5950000
TSCA Yes
Recommended Storage Refrigerator