missing translation for 'onlineSavingsMsg'
Learn More

Butyl Sulfide 98.0+%, TCI America™

Catalog No. p-7118650
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074025ML 25 mL
B0740500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B074025ML Supplier TCI America Supplier No. B074025ML
Only null left

Chemical Identifiers

CAS 544-40-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
IUPAC Name 1-(butylsulfanyl)butane
SMILES CCCCSCCCC

Specifications

Color Colorless
Boiling Point 182°C
Quantity 25 mL
Formula Weight 146.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Sulfide
EINECSNumber (2)-0474
RTECSNumber ER6417000
TSCA Yes